Detector Tubes: Methyl Mercaptan
Actual product may vary
Sensidyne
Detector Tubes: Methyl Mercaptan
Have Questions?
Catalog NumberMeasurement Range
FIS18-071Y5 to 140ppm
Catalog number FIS18-071Y
Price (SGD)
-
Measurement Range:
5 to 140ppm
Detector tubes are sold in packs of 10
Specifications
Components MeasuredMethyl Mercaptan
Measurement Range5 to 140ppm
No. of Strokes1
Quantity10
Response Time1.5 min.
Temperature Range (English)32°F to 104°F
Temperature Range (Metric)0°C to 40°C
Unit SizePack