Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Industrial Hygiene and Environmental Monitoring
▸
Gas and Vapor Detection
▸
Gas Detection Tubes
▸
Detector Tubes: Ethyl Mercaptan
Actual product may vary
Sensidyne
Detector Tubes: Ethyl Mercaptan
Have Questions?
Catalog Number
Measurement Range
FIS18-069H
1 to 160ppm
Catalog number
FIS18-069H
Price (SGD)
-
Measurement Range:
1 to 160ppm
Detector tubes are sold in packs of 10
Specifications
Components Measured
Ethyl Mercaptan
Measurement Range
1 to 160ppm
No. of Strokes
1/2/4
Quantity
10
Response Time
3 min.
Temperature Range (English)
32°F to 104°F
Temperature Range (Metric)
0°C to 40°C