Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Industrial Hygiene and Environmental Monitoring
▸
Gas and Vapor Detection
▸
Gas Detection Tubes
▸
Colorimetric Gas Detector Tubes: Carbonyl Sulphide
Sensidyne
Colorimetric Gas Detector Tubes: Carbonyl Sulphide
Have Questions?
Catalog Number
Measurement Range
FIS18-068L
5 to 60ppm
Catalog number
FIS18-068L
Price (SGD)
-
Measurement Range:
5 to 60ppm
Specifications
Components Measured
Carbonyl Sulfide
Measurement Range
5 to 60ppm
Quantity
2x5 tubes/box
Tube Type
Standard tube