Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Industrial Hygiene and Environmental Monitoring
▸
Gas and Vapor Detection
▸
Gas Detection Tubes
▸
Detector Tubes: Mercaptans (Methyl, Ethyl, Butyl)
Actual product may vary
Sensidyne
Detector Tubes: Mercaptans (Methyl, Ethyl, Butyl)
Have Questions?
Catalog Number
Measurement Range
FIS18-067T
0.5 to 10ppm
Catalog number
FIS18-067T
Price (SGD)
-
Measurement Range:
0.5 to 10ppm
All detector tubes are sold in packs of 10.
Specifications
Components Measured
Mercaptans
Measurement Range
0.5 to 10ppm
No. of Strokes
0.5/1
Quantity
10
Response Time
1 min.
Temperature Range (English)
32°F to 104°F
Temperature Range (Metric)
0°C to 40°C