Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Industrial Hygiene and Environmental Monitoring
▸
Gas and Vapor Detection
▸
Gas Detection Tubes
▸
Detector Tubes: Benzene
Actual product may vary
Sensidyne
Detector Tubes: Benzene
Detector tubes
Have Questions?
Catalog Number
Measurement Range
Response Time
No. of Strokes
FIS18-067J
1 to 100
4 min.
2
Catalog number
FIS18-067J
Price (SGD)
-
Measurement Range:
1 to 100
Response Time:
4 min.
No. of Strokes:
2
All detector tubes are sold in packs of 10.
Specifications
Components Measured
Benzene
Measurement Range
1 to 100
No. of Strokes
2
Quantity
10
Response Time
4 min.
Temperature Range (English)
32°F to 104°F
Temperature Range (Metric)
0°C to 40°C