Replacement Fyrite™ Fluid
Actual product may vary
Bacharach
Replacement Fyrite™ Fluid
For use in Fyrite Gas Analyzer for measuring and analyzing oxygen.
Have Questions?
Catalog NumberFor Use With (Equipment)
FIS23-123026Fyrite Gas Analyzer
Catalog number FIS23-123026
Price (SGD)
-
For Use With (Equipment):
Fyrite Gas Analyzer
  • For Oxygen Range: 0 to 21% and 0 to 60%
  • 3 Bottles/Carton
Specifications
Detection Limits0 to 21% and 0 to 60%
Components MeasuredOxygen
For Use With (Equipment)Fyrite Gas Analyzer
TypeReplacement Fyrite Fluid
For Use With (Application)Measuring and analyzing oxygen
Unit SizePack