Gastec™ Detector Tubes: Hydrogen Sulfide Gas
Actual product may vary
Nextteq
Gastec™ Detector Tubes: Hydrogen Sulfide Gas
Each tube contains a reagent that reacts with a specific gas/vapor, yielding a stain that is measured against calibrations printed on the tube.
Have Questions?
Catalog NumberMeasurement RangeResponse Time
FIS19-026-2940.1 to 4ppm30 sec. to 2 min.
Catalog number FIS19-026-294
Price (SGD)
443.00
Pack
Measurement Range:
0.1 to 4ppm
Response Time:
30 sec. to 2 min.
Price (SGD)
443.00
Pack
Each tube contains a reagent that reacts with a specific gas/vapor, yielding a stain that is measured against calibrations printed on the tube.
Gastec tubes offer accurate, effective readings of more than 500 gases and vapors. Each glass detector tube contains a reagent which reacts with a specific gas or vapor, yielding a stain that is measured against calibrations printed on the tube. The unique absorbing media produce extremely clear lines of stain demarcation for precise reading, and are stable, with a shelf-life of up to two years, depending on the application. (See below for tube selection of specific gases and vapors.) Sold in packs of 10.
Specifications
Components MeasuredHydrogen Sulfide
TypeGas Detection Tube
Measurement Range0.1 to 4ppm
No. of Strokes0.5 to 2
Quantity10
Response Time30 sec. to 2 min.
Temperature Range (English)32°F to 104°F
Temperature Range (Metric)0°C to 40°C
Unit SizePack