Detector Tubes: Water Vapor
Actual product may vary
Sensidyne
Detector Tubes: Water Vapor
Have Questions?
Catalog NumberMeasurement Range
FIS19-0248643 to 80 lb./MMcf
Catalog number FIS19-024864
Price (SGD)
-
Measurement Range:
3 to 80 lb./MMcf
All detector tubes are sold in packs of 10.
Specifications
Components MeasuredWater Vapor
Measurement Range3 to 80 lb./MMcf
Quantity10
Response Time1 min.
Temperature Range (English)32°F to 104°F
Temperature Range (Metric)0°C to 40°C
Unit SizePack