▸
Industrial Hygiene and Environmental Monitoring
▸
Gas and Vapor Detection
▸
Gas Detection Tubes
▸
Colorimetric Gas Detector Tubes: Methyl Mercaptan
Sensidyne
Colorimetric Gas Detector Tubes: Methyl Mercaptan
Have Questions?
Catalog Number
Measurement Range
FIS18-071X
50 to 1000ppm
Catalog number
FIS18-071X
Price (SGD)
-
Measurement Range:
50 to 1000ppm
Specifications
Components Measured
Methyl Mercaptan
Measurement Range
50 to 1000ppm
Quantity
10 Tubes/Box
Temperature Range (English)
32°F to 104°F
Temperature Range (Metric)
0°C to 40°C
Tube Type
Standard tube