Detector Tubes: Hydrogen Sulfide
Actual product may vary
Sensidyne
Detector Tubes: Hydrogen Sulfide
Have Questions?
Catalog NumberMeasurement Range
FIS18-070M1 to 60ppm
Catalog number FIS18-070M
Price (SGD)
-
Measurement Range:
1 to 60ppm
Detector tubes are sold in packs of 10
Specifications
Components MeasuredHydrogen Sulfide
Measurement Range1 to 60ppm
No. of Strokes0.5/1
Quantity10
Response Time1 min.
Temperature Range (English)32°F to 104°F
Temperature Range (Metric)0°C to 40°C
Unit SizePack