Chemware™ Tedlar™ PVF Gas Sampling Bags
Actual product may vary
Saint-Gobain
Chemware™ Tedlar™ PVF Gas Sampling Bags
Good for use with carbon monoxide, sulfur dioxide, hydrogen sulfide, radon and mercaptans
Have Questions?
Catalog NumberCapacity (Metric)Includes
FIS10-923-101.6 LOn/off valve
Catalog number FIS10-923-10
Price (SGD)
-
Capacity (Metric):
1.6 L
Includes:
On/off valve
Can also be used for liquid sampling, when supported with tray and for gas sampling trains.
  • 0.07 in. (2mm) thick PVF
  • Sealed on all four sides
  • Second outboard seal not required
Specifications
Certifications/ComplianceEPA, 40CFR, Part 60, App A Method 18
For Use With (Application)Carbon monoxide, sulfur dioxide, hydrogen sulfide, radon and mercaptans
IncludesOn/off valve
MaterialPVDF
Thickness (English)70 mil
Dimensions (L x W)9 x 9 in. (23 x 23cm)
Length (English)9 in.
Width (English)9 in.
Product TypeGas Sampling Bag
Capacity (English)54 oz.
Capacity (Metric)1.6 L
Length (Metric)23 cm
Width (Metric)23 cm
Thickness (Metric)2 mm
Unit SizeEach