▸
Racks
▸
Special Purpose Racks
▸
Standard Rack for Upright Freezer, 50 mm Box Height
Actual product may vary
Tenak
Standard Rack for Upright Freezer, 50 mm Box Height
Standard Rack fits into any upright freezer and can be used with all standard cryoboxes.
Have Questions?
Catalog Number
Positions
TENTE23136
24
Catalog number
TENTE23136
Price (SGD)
-
Positions:
24
Measure the inside height and depth of your freezer when choosing the right rack
Racks can be used with all freezer brands
Constructed of stainless steel
Specifications
Holds
Boxes
Material
Stainless Steel
Positions
24
For Use With (Equipment)
Upright Freezers
Dimensions (L x W x H)
560 x 139 x 331 mm
Height (Metric)
50 mm (Box)
Array
6 x 4