Nunc™ Seals, Tapes, and Foils
Actual product may vary
Nunc™ Seals, Tapes, and Foils
Nunc™ Seals, Tapes, and Foils
Nunc™ Seals, Tapes, and Foils
Nunc
Nunc™ Seals, Tapes, and Foils
Minimize evaporation and protect samples from contamination and spilling with these sealing tapes, ideal for all assay applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialFor Use With (Application)Certified RNase/DNase-free
NNC236366PolyesterCell culture, sterile applicationsNo
232698AluminumPCR, storage in 96-well platesYes
FIS12-565-398AluminumPCR, storage in 96-well platesYes
FKS10716943AluminumPCR, storage in 96-well platesYes
232701PolypropylenePCRYes
235307PolyolefinPCR, qPCR, ELISA, Protein crystallographyYes
FIS12-565-171VinylLuminescence, photosensitive samplesNo
236366PolyesterCell culture, sterile applicationsNo
FIS12-565-71PolyesterCell culture, sterile applicationsNo
FKS10170671PolyesterCell culture, sterile applicationsNo
NNC236366-PKPolyesterCell culture, sterile applicationsNo
AB-0718PolypropyleneVentilating and storing cell culturesNo
FISAB-0718PolypropyleneVentilating and storing cell culturesNo
FKS10642674PolypropyleneVentilating and storing cell culturesNo
AB-0580PVC-coated PolypropyleneCrystallography and genotypingNo
FKS11524794PVC-coated PolypropyleneCrystallography and genotypingNo
Catalog number NNC236366
Price (SGD)
-
Material:
Polyester
For Use With (Application):
Cell culture, sterile applications
Certified RNase/DNase-free:
No
Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific™ Nunc™ Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.
  • Adhesive seals effectively seal all microplate formats
  • Pressure sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
  • Thin, lightweight tapes seal onto the plate with a convenient, handheld applicator (available separately, Cat. No. 250050)
Specifications
ColorClear
Solvent Or Alcohol ResistantLife Science Reagent Resistant
DescriptionSEALING TAPE POLYESTR STRL CLR
MaterialPolyester
SterilitySterile
AutoclavableNot Autoclavable
No. of Wells96
No. per Case800
No. per Pack200
For Use With (Application)Cell culture, sterile applications
Product TypeAdhesive Seal
Product LineNunc
AdhesiveAcrylic
Certified RNase/DNase-freeNo
PierceableNo
Temperature Range (Metric)-40°C to +120°C
Well ShapeAll well shapes
Plate CompatibilityAll Microplates
Incubation ConditionsGeneral
PeelableYes
Unit SizePack
Contents & Storage
General