0.05% Trypsin-EDTA (1X)
0.05% Trypsin-EDTA (1X)
Actual product may vary
MP Biomedicals
0.05% Trypsin-EDTA (1X)
0.05% Trypsin and 0.5mM EDTA in modified HBSS w/o Ca++ and Mg++, 100 mL
Have Questions?
Catalog NumberQuantity
MPB091689149100 mL
Catalog number MPB091689149
Price (SGD)
93.00
EA
Add to cart
Quantity:
100 mL
Price (SGD)
93.00
EA
Add to cart
Concentration: 0.05% (w/v) Trypsin (1:250) and 0.02% (w/v) EDTA (Versene)
Specifications
For Use With (Application)Cell dissociation, routine cell culture passaging and primary tissue dissociation due to its digestive strength
pH Range7 to 7.4
Sample TypePorcine
FormSolution
Quantity100 mL
Product TypeTrypsin-EDTA
Unit SizeEA
Contents & Storage
0°C to 20°C