▸
Biochemical Reagents
▸
Amino Acids
▸
Glycine, Molecular biology reagent, ≥99%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Glycine, Molecular biology reagent, ≥99%, MP Biomedicals™
Have Questions?
Change view
Quantity:
1 kg
100 g
500 g
5 kg
Catalog number
MPB0219482591
Price (SGD)
150.00
Each
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (SGD)
150.00
Each
Add to cart
Specifications
Chemical Name or Material
Glycine
CAS
56-40-6
Recommended Storage
Store at room temperature (15° to 30°C).
EINECS Number
200-272-2
Assay Percent Range
≥99%
View more
Protein Electrophoresis & Blotting