L-Cysteine [Free Base], MP Biomedicals
L-Cysteine [Free Base], MP Biomedicals
L-Cysteine [Free Base], MP Biomedicals
L-Cysteine [Free Base], MP Biomedicals
MP Biomedicals
L-Cysteine [Free Base], MP Biomedicals
Have Questions?
Catalog NumberQuantity
MPB021946462525 g
Catalog number MPB0219464625
Price (SGD)
149.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (SGD)
149.00
EA
Add to cart
Specifications
Chemical Name or MaterialL-Cysteine [Free Base]
CAS52-90-4
Recommended StorageRoom temperature
Molecular FormulaC3H7NO2S
Synonyml-cysteine, cysteine, cystein, half-cystine, r-cysteine, thioserine, r-2-amino-3-mercaptopropanoic acid, l-+-cysteine, l-cystein, 2r-2-amino-3-sulfanylpropanoic acid
View more