Kimtech™ Sterling™ Nitrile-XTRA 12 in. Gloves
Actual product may vary
Kimtech™ Sterling™ Nitrile-XTRA 12 in. Gloves
Sterling™ Nitrile-XTRA gloves are 12 in. in length for added protection and combine the sensitivity of latex with the strength of nitrile.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
KLC53140Large
KLC53137X-Small
FIS19-050-591Small
KLC53138Small
FIS19-050-592Medium
KLC53139Medium
Catalog number KLC53140
Price (SGD)
320.00
Special offer
Ends: 06-Sep-2026
383.00
Save 63.00 (16%)
Case
Size:
Large
Price (SGD)
320.00
Special offer
Ends: 06-Sep-2026
383.00
Save 63.00 (16%)
Case
Sterling™ Nitrile-XTRA gloves are 12 in. in length for added protection and combine the sensitivity of latex with the strength of nitrile.
  • XTRA long 12 in. length
  • Powder free
  • non-sterile
  • Static dissipative
  • Textured fingertips
  • Beaded cuff
  • Ambidextrous
  • Typical uses include: Laboratory, Cleaning, Maintenance, Electronic Assembly and Inspection
Specifications
Certifications/ComplianceASTM D 6319, ASTM D6978-05, Meets NFPA standard 1999:2013, Meets USP <800> guidelines
Chemotherapeutic RatedYes
ApplicationsHealthcare, Laboratory, Industrial
AQL1
ColorGray
DisposableSingle-use
Exterior FinishPartially Textured
Finger Thickness (English)3.5 mil
Finger Thickness (Metric)0.08 mm
Latex FreeYes
MaterialNitrile
Palm Thickness (English)3.1 mil
Palm Thickness (Metric)0.07 mm
PowderedPowder Free
SizeLarge
Thickness (English)3.5 mil
Thickness (Metric)0.08 mm
FDA 510 (k)Yes
Model53140
SeriesKimtech™
Tensile Strength42 MPa
Length (English)12 in.
Length (Metric)30.5 cm
Unit SizeCase