Specifications
Certifications/ComplianceASTM D5151
Chemotherapeutic RatedNo
ApplicationsHealthcare, Laboratory
AQL1.5
ColorPurple
DisposableSingle-use
Exterior FinishPartially Textured
Finger Thickness (English)2.8 mil
Finger Thickness (Metric)0.07 mm
Latex FreeYes
MaterialNitrile
Palm Thickness (English)2 mil
Palm Thickness (Metric)0.05 mm
PowderedPowder Free
SizeX-Small
Thickness (English)2.8 mil
Thickness (Metric)0.07 mm
FDA 510 (k)Yes
Model52816
Tensile Strength24 MPa
Length (English)9.5 in.
Length (Metric)24.13 cm
Unit SizeCase