Purple Nitrile™ Ambidextrous Gloves
Actual product may vary
Purple Nitrile™ Ambidextrous Gloves
Purple Nitrile™ Ambidextrous Gloves
Purple Nitrile™ Ambidextrous Gloves
Purple Nitrile™ Ambidextrous Gloves
Purple Nitrile™ Ambidextrous Gloves
Fisherbrand
Purple Nitrile™ Ambidextrous Gloves
Offering the highest protection of all Kimtech™ gloves, Purple Nitrile™ gloves are latex-, silicone- and powder-free. Textured fingertips and beaded cuffs ensure strength and sensitivity, and the gloves are also anti-static and food contact approved.
Have Questions?
Catalog NumberSize
FKS11889610Medium
Catalog number FKS11889610
Price (SGD)
-
Size:
Medium
Assured Compliance:
  • PPE Cat III according to Regulation (EU) 2016/425
  • EN ISO 374-1:2016 Type B (JKT) Chemical Splash protection
  • EN 374-4:2003 Resistance to degradation by chemicals
  • EN ISO 374-5:2016 Micro Organism and VIRUS Protection
  • Food contact approved
  • Industry-leading gloves offering unrivalled protection, cleanliness and quality.
  • Nitrile construction results in products that are stronger and leaner than latex gloves, and feature better protection against a wider range of chemicals, including cytotoxic drugs.
  • Gloves are anti-static tested to protect the wearer and equipment, and ambidextrous.
  • Textured fingertips enhance grip and tactile sensitivity for safer and more efficient processes.
  • Beaded cuffs add strength to the gloves, reducing the risk of tearing and increasing their durability, while also reducing roll down for easier donning and doffing.
  • Contain no natural rubber latex, silicone or powder, reducing the risks of skin irritation for the wearer.
Specifications
Certifications/CompliancePPE Category III by EC Council Directive 89/686/EEC. Tested in accordance with the EN Norms EN420:2003, ISO 16604 procedure B:2004, EN1149-5:2003, EN421:2004, Meets or exceeds AQL 0.65 G1 Inspection for pinholes, ISO 9001 and ISO 13485, FDA CFR 21 part 820
ClassificationPPE Category III
Accelerator FreeYes
ColorPurple
DisposableSingle-use
Exterior FinishSmooth
Finger Thickness (English)6.3 mil
Finger Thickness (Metric)0.16 mm
HandAmbidextrous
Latex FreeYes
MaterialNitrile
Palm Thickness (English)5.5 mil
Palm Thickness (Metric)0.14 mm
PowderedPowder Free
Product TypeNitrile Examination Glove
SizeMedium
SterilityNon-sterile
Thickness (English)5.5 mil
Thickness (Metric)0.09 mm (Cuff)
Finger StyleTextured
Inspected for Medical UseYes
Quantity100 Pk.
Resistant ToPuncture and abrasion
Tensile Strength21 MPa
Length (English)9.6 in.
Length (Metric)24.4 cm
Unit SizePack