Clear Polystyrene 96-Well Microplates
Actual product may vary
Fisherbrand
Clear Polystyrene 96-Well Microplates
Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ Clear Polystyrene 96-Well Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 96-well microplates for laboratory miniaturization and automation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWell ShapeSurface TreatmentSterilityLid
FKS10296381RoundNon-treatedSterileWithout Lid
FIS07-200-107Conical Bottom (V-well)Non-treatedSterileWithout Lid
FIS07-200-108Conical Bottom (V-well)Non-treatedNon-sterileWithout Lid
CRG3370Flat Bottom (F-well)Non-treatedSterileWith Lid
Catalog number FKS10296381
Price (SGD)
835.00
Pack
Well Shape:
Round
Surface Treatment:
Non-treated
Sterility:
Sterile
Lid:
Without Lid
Price (SGD)
835.00
Pack
  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
Specifications
FormatStandard
LidWithout Lid
Array8 x 12
PackagingIndividually Wrapped
Binding TypeNot Treated
ColorClear
Product TypeEIA/RIA
MaterialPolystyrene
No. of Wells96
Volume (Metric) Working75 to 200 μL
SterilitySterile
Surface TreatmentNon-treated
Well ShapeRound
For Use With (Application)Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions.
No. per Case50/Cs.
No. per Pack1/Pk.
Unit SizePack