Abgene™ 96 Well 1.2 mL Polypropylene DeepWell™ Sample Processing & Storage Plate
Actual product may vary
Abgene™ 96 Well 1.2 mL Polypropylene DeepWell™ Sample Processing & Storage Plate
Abgene™ 96 Well 1.2 mL Polypropylene DeepWell™ Sample Processing & Storage Plate
Abgene™ 96 Well 1.2 mL Polypropylene DeepWell™ Sample Processing & Storage Plate
Abgene™ 96 Well 1.2 mL Polypropylene DeepWell™ Sample Processing & Storage Plate
Thermo Scientific
Abgene™ 96 Well 1.2 mL Polypropylene DeepWell™ Sample Processing & Storage Plate
Optimize suspension cell culture with these U-bottomed 1.2 mL storage plates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWell ShapePackaging
FISAB-1127Square, Round Bottom (U-well)Individually Wrapped
AB-1127Square, Round Bottom (U-well)Individually Wrapped
FKS10243223Square, Round Bottom (U-well)Individually Wrapped
AB-0564RoundIndividually Wrapped
FKS12114172RoundIndividually Wrapped
FISAB-0787RoundBulk
Catalog number FISAB-1127
Price (SGD)
1,091.00
Case
Well Shape:
Square, Round Bottom (U-well)
Packaging:
Individually Wrapped
Price (SGD)
1,091.00
Case
Thermo Scientific™ Abgene™ polypropylene microplates and deepwell plates offer storage security for assays, compound libraries or storing samples for either intermediate or long term use. Abgene microtiter plates are manufactured to exacting specifications in our Class 100,000 clean room ISO 9001 conditions using high quality medical grade virgin polypropylene resins, which ensure confidence in the quality and performance of the storage plates. Designed to ANSI standards these microplates and deepwell plates are compatible with a variety of automated liquid handling for high throughput workflows. The polypropylene storage plates are offered with spindle, pyramidal, U and V bottom wells along with a wide range of volumes to accommodate most applications.
Features
  • Round well shape designed for optimal sample recovery or square shape to maximize sample volume within ANSI footprint design
  • Each well has an independent sealing rim to prevent cross-contamination
  • U-bottomed wells ideally suited for sample resuspension
  • Polypropylene (AB-1068, polystyrene)
  • Resistant to DMSO (100%); Ethanol (100%); Isopropanol (100%)
  • Suitable for Autoclaving (15 minutes at 121°C) or Gamma Irradiation
  • Sealing options: Adhesive Seals, Heat Seals, Storage Plate Caps and Cap Strips, and Storage Plate Sealing Mats
  • Cleanroom manufactured
  • ANSI-format for compatibility with automated systems
Specifications
BarcodeCustom Barcoding Available
Certifications/ComplianceCertified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant
FormatStandard
Description96-well Polypropylene Sample Processing & Storage Plates
Array8 x 12
AutoclavableAutoclavable
PackagingIndividually Wrapped
Binding TypeLow binding PP
Product LineAbgene
ColorNatural
Product Type96-well plate
For Use With (Equipment)ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
MaterialPolypropylene
Volume (Metric) Well1200 μL
No. of Wells96
Volume (Metric) Working1000 μL
SterilitySterile
Surface TreatmentNon-treated
Well ShapeSquare, Round Bottom (U-well)
Dnase FreeYes
For Use With (Application)Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
Max. RCF2000 x g
No. per Case50
No. per Pack1
Regulatory StatusRUO
RNase FreeYes
Fit for PurposePCR Ready
Plate SizeANSI/ SLAS Standard
Unit SizeCase