Immuno Clear Standard Modules
Actual product may vary
Immuno Clear Standard Modules
Immuno Clear Standard Modules
Immuno Clear Standard Modules
Thermo Scientific
Immuno Clear Standard Modules
Perform applications that deliver well-to-well reproducibility with these immuno clear standard modules. Manufactured to fit standard equipment.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSurface TreatmentVolume (Metric) WorkingWell ShapeWell Design
FIS22-047029MaxiSorp350 μLRound, Flat Bottom (F-well)F8
468667MaxiSorp350 μLRound, Flat Bottom (F-well)F8
469078PolySorp350 μLRound, Flat Bottom (F-well)F8
469914MaxiSorp350 μLRound, Flat Bottom (F-well)F16 (2x8 wells)
469949MaxiSorp350 μLRound, Flat Bottom (F-well)F8
475078MaxiSorp250 μLRoundU8
95029100Enhanced Binding350 μLRound, Flat Bottom (F-well)F8
FIS21-377-254Enhanced Binding350 μLRound, Flat Bottom (F-well)F8
Catalog number FIS22-047029
Price (SGD)
4,499.00
Case
Surface Treatment:
MaxiSorp
Volume (Metric) Working:
350 μL
Well Shape:
Round, Flat Bottom (F-well)
Well Design:
F8
Price (SGD)
4,499.00
Case
Perform applications which are delivering well-to-well reproducibility with Thermo Scientific™ Immuno Clear Standard Modules, which feature Nunc™, Immulon™ and Microtiter™ module plates developed for ELISA techniques. Manufactured to fit standard equipment, modules provide flexible use of components with small sample sizes. They are backed by industry-leading quality standards and certifications.
Features:
  • Several strip formats: Eight-well strip, 12-well strip, 16-well strip (two columns of eight wells)
  • Several well shapes: flat bottom, C bottom (radius edge), U shape wells, StarWells (wells have fins to increase the surface area and enhance performance)
  • Several surfaces to maximize assay performance
  • Modules that remain in frame when inverted
  • Stackable frame marked for lid orientation
  • Polystyrene wells in polyethylene frame
  • Ext. Dim.: 128 × 86mm
Choice of Surface:
  • Thermo Scientific™ PolySorp™, Immulon™ 1B, and Universal Binding (UB) are for adsorption of hydrophobic molecules.
  • Thermo Scientific™ Medisorp™ and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules.
  • Thermo Scientific™ MaxiSorp™, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays.
  • Thermo Scientific™ MultiSorp™ are very hydrophilic and works well for glycans and water soluble proteins.
Ordering Information:
Modules are freestanding. Cat. No. 95029440 and 464394 are certified. Frames and modules are available separately. Sealing tape and strip caps are available. Color-coded modules are available on request. Contact your customer service or sales representative for further details.
Specifications
BreakapartNo
Binding PropertyHydrophilic
Certifications/ComplianceCertified for adsorption of IgG; CV of <5% between wells
CertifiedYes
FormatStandard
LidWithout Lid
Detection MethodColorimetric
Array8 x 12
Packaging120 Cases
Binding TypeAdsorption
ColorClear
Product TypeMicroplate Strip
MaterialPolystyrene
No. of Wells96
Volume (Metric) Working350 μL
SterilityNon-sterile
Surface TreatmentMaxiSorp
Well ShapeRound, Flat Bottom (F-well)
No. per Case120
No. per Pack20
Plate BlockingNone
Quantity120/Cs.
Target MoleculeAntibodies (Glycoproteins)
Well DesignF8
Unit SizeCase