MICROFLEX™ XCEED™ XC-310 Nitrile Gloves
Actual product may vary
Microflex
MICROFLEX™ XCEED™ XC-310 Nitrile Gloves
Ergonomically designed to reduce hand fatigue
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS19-177-523Large
FIS19-177-521Small
FIS19-177-522Medium
Catalog number FIS19-177-523
Price (SGD)
71.00
Pack
Size:
Large
Price (SGD)
71.00
Pack
Thin, powder free nitrile exam glove with ERGOFORM™ is designed to reduce hand fatigue.
  • Reduced allergy risk: MICROFLEX™ XCEED™ XC-310 gloves are manufactured without accelerators for low dermatitis potential. These gloves prevent instances of Type I latex allergies and Type IV chemical allergic reactions
  • Extra durability: stronger formulation to protect against rips and tears while keeping the same tactile sensitivity
  • Enhanced comfort: softer formulation for a lightweight, softer skin feel
  • ERGOFORM™ Technology: MICROFLEX™ XCEED™ XC-310 gloves are designed with proprietary ergonomic design technology to support musculoskeletal health and increase worker productivity
  • Reduced waste: Gloves are smartly packaged, with 250 gloves per box and easy, single glove dispensing to reduce waste
  • Strong barrier integrity: With a low 0.65 AQL, these gloves offer advanced barrier integrity
  • Specialized protection: These medical exam gloves are specially tested against fentanyl to ensure advanced protection and peace of mind
Key Features and Benefits
  • Made without accelerators: To protect from allergic reactions
  • ERGOFORM™ Technology: Ergonomic design supports musculoskeletal health
  • Fentanyl-tested: For enhanced user protection
Specifications
Certifications/ComplianceASTM D6319, ASTM D6978, Food Type: Rubber, EN ISO 21420:2020, FDA21 CFR 177-2600-US Food Contact Approved, US QSR/FDA 510(k) Medical Examination Grade
Certifications/Compliance (Europe)EN 1149-1/2/3
Chemotherapeutic RatedYes
ChlorinatedYes
Accelerator FreeYes
ApplicationsIndustrial, Healthcare, Laboratory
AQL0.65
ColorBlue
DisposableSingle-use
Exterior FinishPartially Textured
Finger Thickness (English)4.3 mil
Finger Thickness (Metric)0.11 mm
Latex FreeYes
MaterialNitrile
Palm Thickness (English)2.8 mil
Palm Thickness (Metric)0.07 mm
PowderedPowder Free
Product TypeNitrile Examination Glove
SizeLarge
SterilityNon-sterile
Thickness (English)3 mil
Thickness (Metric)0.07 mm
FDA 510 (k)Yes
Glove Cuff StyleBeaded
ModelXC-310
SeriesMICROFLEX™ XCEED™ XC-310
Tensile Strength33 MPa
Length (English)9.5 in.
Length (Metric)24 cm
Unit SizePack