Colormetric Carbon Replacement Cartridge for Coalescing/Carbon Filters
Actual product may vary
Justrite
Colormetric Carbon Replacement Cartridge for Coalescing/Carbon Filters
Replacement cartridge for use with Coalescing/Carbon Filters 28161 and 28162.
Have Questions?
Catalog NumberFor Use With (Equipment)
FIS19-052-374Justrite Coalescing/Carbon Filters 28161/28162
Catalog number FIS19-052-374
Price (SGD)
892.00
Each
For Use With (Equipment):
Justrite Coalescing/Carbon Filters 28161/28162
Price (SGD)
892.00
Each
  • Color-changing carbon cartridge (top portion) changes from purple to black as hydrocarbons are absorbed, indicating when replacement is necessary
  • Coalescing filter (bottom portion) can be reused with new carbon filters several times and replaced after 3-4 carbon filter cycles
Specifications
Certifications/ComplianceEPA 40 CFR 264.173
Product TypeReplacement Cartridge for Coalescing Carbon Filters
For Use With (Application)HPLC
For Use With (Equipment)Justrite Coalescing/Carbon Filters 28161/28162
MaterialPlastic
Width (English)4 in.
Width (Metric)10.16 cm
Height (English)11 in.
Height (Metric)27.94 cm
Unit SizeEach