Specifications
Product TypeCleanroom Wipe
Length (English)9 in.
Length (Metric)22.86 cm
Width (English)9 in.
Width (Metric)22.86 cm
MaterialPolyester, Rayon
CompatibilityCommon cleaning and disinfecting solutions
AutoclavableNot Autoclavable
Basis Weight61.9g/m3
ColorWhite
ConstructionHydroentangled nonwoven blend
Dimensions (L x W)9 x 9 in. (22.86 x 22.86 cm)
For Use With (Application)Cleaning of lab equipment
Packaging Quantity300 Sheets/Pk.
Unit SizePack