MICROFLEX™ Diamond Grip™ Plus DGP-350 Latex Gloves
Actual product may vary
Microflex
MICROFLEX™ Diamond Grip™ Plus DGP-350 Latex Gloves
Multi-purpose gloves feature a thick durable design and are fully textured for use in a variety of industries including healthcare, life sciences and food processing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS19-050-080CMedium
FIS19-050-080AX-Small
FIS19-050-080BSmall
Catalog number FIS19-050-080C
Price (SGD)
39.00
Pack
Size:
Medium
Price (SGD)
39.00
Pack
  • Natural rubber latex construction offers high elasticity for greater comfort and less hand fatigue
  • Tactile sensitivity
  • Fully textured exterior ensures secure grip
  • Tapered fingers for greater comfort
  • Beaded cuffs for easy donning and doffing
  • Powder free, non-sterile
  • Ambidextrous, single-use
  • Thickness: 6.3 mil fingertips
  • Length: 9.6 in. (24.5 cm)
  • Natural color
Specifications
Certifications/ComplianceASTM D3578, FDA21 CFR 177-2600
Certifications/Compliance (Europe)EN 420:2003 + A1:2009
Chemotherapeutic RatedNo
ApplicationsHealthcare, Laboratory, Industrial
AQL1.5
ColorNatural
DisposableSingle-use
Exterior FinishFully Textured
Finger Thickness (English)6.3 mil
Finger Thickness (Metric)0.16 mm
Laboratory/Cleanroom UseNo
Latex FreeNo
MaterialLatex
Palm Thickness (English)5.1 mil
Palm Thickness (Metric)0.13 mm
PowderedPowder Free
Product TypeLatex Examination Gloves
SizeMedium
SterilityNon-sterile
Thickness (English)6.3 mil
Thickness (Metric)0.16 mm
FDA 510 (k)Yes
Glove Cuff StyleBeaded
ModelDGP350-M
SeriesDiamond Grip Plus™ DGP-350
Tensile Strength22 MPa
Length (English)9.5 in.
Length (Metric)24.1 cm
Product LineMICROFLEX Diamond Grip Plus
Unit SizePack