MicroSeal™ 1200 Wipers
Actual product may vary
Berkshire
MicroSeal™ 1200 Wipers
For ISO Class 3 (Class 1) applications
Have Questions?
Catalog NumberDimensions (L x W)
FIS19-037-6039 x 9 in. (22.86 x 22.86 cm)
Catalog number FIS19-037-603
Price (SGD)
193.00
Pack
Dimensions (L x W):
9 x 9 in. (22.86 x 22.86 cm)
Price (SGD)
193.00
Pack
All-purpose wipers for use in spill control, equipment cleaning, and general cleanup.
  • Made of ultrasonically sealed edge 100% continuous filament polyester knit
  • Works well with IPA, acetone, and other solvents
  • Laundered and packed in an ISO 4 (Class 10) cleanroom
  • Critically low particle and fiber generation
Specifications
Product TypeCleanroom Wipe
Length (English)9 in.
Length (Metric)22.86 cm
Width (English)9 in.
Width (Metric)22.86 cm
MaterialPolyester Knit
CompatibilityIPA, Acetone and other solvents
EdgesSealed
Dimensions (L x W)9 x 9 in. (22.86 x 22.86 cm)
For Use With (Application)For use in ISO Class 3, spill control, equipment cleaning and general cleanup
For Use With (Equipment)Photolithography Process areas, flat panel, optics, and chamber cleans
Unit SizePack