Spill-Control Drum Funnel
Actual product may vary
Spill-Control Drum Funnel
Spill-Control Drum Funnel
Eagle
Spill-Control Drum Funnel
Spout fits into any 2 in. (5.1cm) NPT bung
Have Questions?
Catalog NumberProduct Type
FIS18-600Drum Funnel
Catalog number FIS18-600
Price (SGD)
194.00
Each
Product Type:
Drum Funnel
Price (SGD)
194.00
Each
Polyethylene funnel fits 55 gal. (208L) and 30 gal. (114L) tight-head drums; also fit most 5 gal. (19L) pails.
  • High sidewall holds 5 gal. (19L) of liquid
  • Available with or without brass screen
  • Optional funnel cover (cat. no. 18884B) keeps out dirt and contaminants
Brass screen is not a flame arrester.
Specifications
Certifications/ComplianceU.S. EPA 40CFR265.173
Product TypeDrum Funnel
Capacity (Metric)19 L
ColorYellow
For Use With (Application)Spill Control
For Use With (Equipment)55 and 30 gal. tight-head drums, 5 gal. pails
MaterialHDPE
QuantityEa.
Height (Metric)127 mm
Diameter (Metric) Opening460 mm
Unit SizeEach