Polyester Sealed-Edge Cleanroom Wipes
Actual product may vary
Fisherbrand
Polyester Sealed-Edge Cleanroom Wipes
Polyester Sealed-Edge Cleanroom Wipes are absorbent and extremely clean, designed for ultra-critical environments. They contain laser cut, heat sealed-edges and constructed from double knit 100% no-run polyester.
Have Questions?
Catalog NumberDimensionsLxW
FIS17-444-001
Catalog number FIS17-444-001
Price (SGD)
163.00
Pack
Price (SGD)
163.00
Pack
Polyester Sealed-Edge Cleanroom Wipes are ultra-clean wipes ideal for critical cleanrooms (ISO Class 3-4 and higher). They are made from made from double knit 100% polyester with laser cut, heat sealed-edges. These polyester wipes are double-bagged, cleanroom packaged, and manufactured in a cleanroom environment. Wipes are manufactured and processed to meet the demands of the most critical environments. Made from the highest quality, lowest-linting substrates - free of bonding agents, adhesives or chemical additives.
Cleanroom Compatibility
  • ISO Class 3-8 (Class 1-100,000) Cleanrooms
Features
  • Manufactured from continuous run 100% polyester providing a tough, absorbent and abrasion resistant product
  • Production process guarantees extremely low particle generation and chemical extractables
  • Compatible with virtually any solvent or solution for cleaning and wiping
  • Double bagged and vacuum cleanroom packed
  • Provides lot-to-lot traceability
Specifications
Product TypeCleanroom Wipe
Length (English)12 in.
Length (Metric)30.48 cm
Width (English)12 in.
Width (Metric)30.48 cm
MaterialPolyester
Absorbancy310 mL/m2
AutoclavableNot Autoclavable
Basis Weight140 gsm
Certifications/ComplianceISO Class 3-8 class 1-10 cleanrooms, Meets USP 797 and USP 800 wipe requirements
ColorWhite
For Use With (Application)Laboratory and Cleanroom
Packaging Quantity150 Wipes/Bag, 10 Bags/Cs.