16-Position Cuvette Rack
Actual product may vary
BrandTech
16-Position Cuvette Rack
Convenient storage for standard cuvettes.
Have Questions?
Catalog NumberHolds
FIS14-377-020Cuvettes (10 mm)
Catalog number FIS14-377-020
Price (SGD)
153.00
Each
Holds:
Cuvettes (10 mm)
Price (SGD)
153.00
Each
  • Holds 16 standard-sized cuvettes in numbered positions
  • Autoclavable to 121°C (249.8°F)
Specifications
AutoclavableAutoclavable
ColorGray
HoldsCuvettes (10 mm)
MaterialPolypropylene
Positions16
ShapeRectangular
StackableNo
For Use With (Equipment)Cuvettes
Length (Metric)210 mm
Width (English)2.8 in.
Height (English)1.5 in.
Product TypeCuvette Rack
Height (Metric)38 mm
Width (Metric)70 mm
Unit SizeEach