Storage Microplates
Actual product may vary
Axygen
Storage Microplates
For sample collection, in-vitro growth chambers and long term storage.
Have Questions?
Catalog NumberColor
FIS14-222-241Clear
Catalog number FIS14-222-241
Price (SGD)
92.00
Pack
Color:
Clear
Price (SGD)
92.00
Pack
Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.
  • 450 μL deep well plate
  • ANSI/SLAS 1-2004: Microplates - Footprint Dimensions
  • Ultra-flat surface ensures proper sealing with heat sealing film
Specifications
Certifications/ComplianceANSI, SLAS 1-2004
LidWithout Lid
Array8 x 12
AutoclavableAutoclavable
ColorClear
Product Type96-well Storage V-bottom Microplate
MaterialPolypropylene
No. of Wells96
SterilitySterile
Well ShapeConical Bottom (V-well)
For Use With (Application)Sample collection, in-vitro growth chambers and long term storage
Max. RCF4000 x g
No. per Case50/Cs.
No. per Pack10/Pk.
Temperature Range (English)-112°F
Temperature Range (Metric)-80°C
Unit SizePack