MICROFLEX™ EVOLUTION ONE™ EV-2050 Latex Exam Gloves
Actual product may vary
Fisherbrand
MICROFLEX™ EVOLUTION ONE™ EV-2050 Latex Exam Gloves
Exam gloves provide protection with maximum comfort for long shifts.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FIS11-462-68BSmall
FIS11-462-68AX-Small
FIS11-462-68CMedium
FIS11-462-68DLarge
Catalog number FIS11-462-68B
Price (SGD)
45.00
Pack
Size:
Small
Price (SGD)
45.00
Pack
  • Comfortable latex: Soft latex provides comfort during extended wear
  • Fully textured: Offering a secure grip
  • Food handling compliance: Suitable for food handling applications
N/A
Specifications
Certifications/ComplianceASTM D3578, FDA21 CFR 177-2600
Certifications/Compliance (Europe)EN 420:2003 + A1:2009
Chemotherapeutic RatedNo
ChlorinatedYes
Accelerator FreeNo
ApplicationsLaboratory, Healthcare
AQL1.5
ColorNatural
DisposableReusable
Exterior FinishFully Textured
Finger Thickness (English)5.9 mil
Finger Thickness (Metric)0.15 mm
Laboratory/Cleanroom UseNo
Latex FreeNo
MaterialLatex
Palm Thickness (English)5.5 mil
Palm Thickness (Metric)0.14 mm
PowderedPowder Free
Product TypeExamination Gloves
SizeSmall
SterilityNon-sterile
Thickness (English)5.9 mil
Thickness (Metric)0.15 mm
FDA 510 (k)Yes
Glove Cuff StyleBeaded
ModelEV-2050
SeriesEvolution One™ EV-2050
Tensile Strength25 MPa
Length (English)9.6 in.
Length (Metric)24.4 cm
Unit SizePack