▸
Microplates
▸
Microplate Covers
▸
Microplate Sealing Films and Tapes
▸
Adhesive Plate Seals
Actual product may vary
Adhesive Plate Seals
Seals are optically clear for PCR applications
Have Questions?
Catalog Number
Material
FIS08-408-240
Polypropylene
Catalog number
FIS08-408-240
Price (SGD)
183.00
Case
Add to cart
Material:
Polypropylene
Price (SGD)
183.00
Case
Add to cart
Seals most plates
Rated from -40 to ±125°C
Polypropylene
Specifications
Color
Clear
Material
Polypropylene
Sterility
Non-sterile
No. per Case
100
For Use With (Application)
For PCR applications
Product Type
Plate Seal
Temperature Range (Metric)
-40°C to +125°C