Small Volume™ 384 Well HiBase Polypropylene Microplates
Actual product may vary
Greiner Bio-One
Small Volume™ 384 Well HiBase Polypropylene Microplates
Reduces dead volume. Greiner Bio-One Small Volume™ 384 Deep Well Polypropylene Microplates are specifically designed to accommodate heat-sealing systems.
Have Questions?
Catalog NumberColor
FIS07-000-916Black
Catalog number FIS07-000-916
Price (SGD)
1,424.00
Case
Color:
Black
Price (SGD)
1,424.00
Case
  • Ultra low non-specific biomolecular binding properties (proteins, DNA, RNA)
  • Long-term surface stability without degradation or leaching
  • Higher assay sensitivity with reduced background
  • Good signal to noise ratio
  • Fre
Specifications
BarcodeAlphanumeric
Array16 x 24
AutoclavableNot Autoclavable
PackagingInner packs of 10
ColorBlack
MaterialPolystyrene
No. of Wells384
SterilityNon-sterile
Surface TreatmentProtein-Repellent Non-binding
Total Volume (Metric) Well28 μL
Well ShapeFlat Bottom (F-well)
Quantity40/Cs.
Shelf Life4 Years after month of production
Temperature Range (Metric)-20°C to +60°C
Well ColorBlack
Unit SizeCase