UV-Star™ 96-well UV Spectroscopy Microplate
Actual product may vary
Greiner Bio-One
UV-Star™ 96-well UV Spectroscopy Microplate
For use in UV Spectroscopy. Greiner Bio-One UV-Star™ 96-Well UV Spectroscopy Microplate is used for measurements of protein concentration at 280nm.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialWell Design
FIS07-000-407Cycloolefin Copolymer, Cycloolefin CopolymerChimney
FIS07-000-724PolystyreneHalf area
Catalog number FIS07-000-407
Price (SGD)
1,238.00
Case
Material:
Cycloolefin Copolymer, Cycloolefin Copolymer
Well Design:
Chimney
Price (SGD)
1,238.00
Case
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Can be traced back to production through defined LOT number system
  • Optical window down to 230nm ideal for nucleic acid
Specifications
BreakapartNo
FormatStandard
LidWithout Lid
Array8 x 12
PackagingInner bags of 10
Binding TypeNon Binding
ColorClear
Product Type96-well Microplate
MaterialCycloolefin Copolymer, Cycloolefin Copolymer
No. of Wells96
SterilityNon-sterile
Well ShapeFlat Bottom (F-well)
Dnase FreeYes
For Use With (Application)UV Spectroscopy
Quantity40/Cs.
Well DesignChimney
Dimensions (L x W x H)127.8 x 85.5 x 11.5 mm
Unit SizeCase