Ultra Low Profile Polystyrene Microplate Lids
Actual product may vary
Ultra Low Profile Polystyrene Microplate Lids
Ultra Low Profile Polystyrene Microplate Lids
Greiner Bio-One
Ultra Low Profile Polystyrene Microplate Lids
Used for high-format low profile microplates
Have Questions?
Catalog NumberSterility
FIS07-000-297Non-sterile
Catalog number FIS07-000-297
Price (SGD)
780.00
Case
Sterility:
Non-sterile
Price (SGD)
780.00
Case
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic, non-cytotoxic
  • Offer protection against contamination and evaporation during sample storage and cell cultivation
  • Compatible with a range of different microplates
For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.
Specifications
ColorClear
Dimensions (L x W x H)128 x 85 x 6 mm
MaterialPolystyrene
SterilityNon-sterile
Condensation RingsNo
No. per Case100
No. per Pack5
Plate CompatibilityA variety of microplates, particularly well suited for high-format, low-profile microplates
Unit SizeCase