CELLCOAT™ μClear™ 96-well, Poly-D Lysine-Treated, Flat-Bottom Microplate
Actual product may vary
Greiner Bio-One
CELLCOAT™ μClear™ 96-well, Poly-D Lysine-Treated, Flat-Bottom Microplate
Color:Black, Clear Bottom; Sterility:Non-sterile; Lid:With Lid
Have Questions?
Catalog NumberColor
FIS07-000-190Black, Clear Bottom
Catalog number FIS07-000-190
Price (SGD)
1,763.00
Case
Color:
Black, Clear Bottom
Price (SGD)
1,763.00
Case
  • Reduced-serum or serum-free cultivation
  • Cell differentiation and neuron growth
  • Promotion of cell adhesion, proliferation and growth of transfected cell lines
  • Increase in isolation and cultivation efficiency
Specifications
LidWith Lid
Array8 x 12
AutoclavableNot Autoclavable
ColorBlack, Clear Bottom
MaterialPolystyrene
No. of Wells96
Volume (Metric) Working25 to 340 μL
SterilityNon-sterile
Surface TreatmentPoly-D-Lysine
Total Volume (Metric) Well392 μL
Well ShapeFlat Bottom (F-well)
Max. RCF4800 x g (swinging bucket rotor)
No. per Case20
No. per Pack5
Quantity20/Cs.
Temperature Range (Metric)Room Temperature
Well DesignChimney
Unit SizeCase