CELLCOAT™ 96-well, Collagen Type I-Treated, Flat-Bottom Microplate
Actual product may vary
Greiner Bio-One
CELLCOAT™ 96-well, Collagen Type I-Treated, Flat-Bottom Microplate
Color:Black; Sterility:Non-sterile; Lid:With Lid
Have Questions?
Catalog NumberColor
FIS07-000-186Black
Catalog number FIS07-000-186
Price (SGD)
1,869.00
Case
Color:
Black
Price (SGD)
1,869.00
Case
  • Cell cultivation in serum-free or serum-reduced medium
  • Rat tail Collagen Type I
  • Increase in isolation and cultivation efficiency
  • Immediate use, time-saving
  • Consistent quality
  • Storable at room temperature
Specifications
LidWith Lid
Array8 x 12
AutoclavableNot Autoclavable
ColorBlack
MaterialPolystyrene
No. of Wells96
Volume (Metric) Working25 to 340 μL
SterilityNon-sterile
Surface TreatmentCollagen Type I
Total Volume (Metric) Well392 μL
Well ShapeFlat Bottom (F-well)
Max. RCF4800 x g (swinging bucket rotor)
No. per Case20
No. per Pack5
Quantity20/Cs.
Temperature Range (Metric)Room Temperature
Well DesignChimney
Unit SizeCase