CELLSTAR 96-well, Cell Culture-Treated, U-Shaped-Bottom Microplate
Actual product may vary
Greiner Bio-One
CELLSTAR 96-well, Cell Culture-Treated, U-Shaped-Bottom Microplate
Color:Clear; Sterility:Sterile; Lid:With Lid
Have Questions?
Catalog NumberColor
FIS07-000-164Clear
Catalog number FIS07-000-164
Price (SGD)
979.00
Case
Color:
Clear
Price (SGD)
979.00
Case
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Non-Cytotoxic
  • Compatible with automated systems
  • Alphanumeric well coding
  • Conforms to recommendation of American National Standards Institute (ANSI) (A
Specifications
LidWith Lid
Array8 x 12
AutoclavableNot Autoclavable
ColorClear
MaterialPolystyrene
No. of Wells96
Volume (Metric) Working40 to 280 μL
SterilitySterile
Surface TreatmentCell Culture
Total Volume (Metric) Well323 μL
Well ShapeRound Bottom (U-well)
Max. RCF1000 x g (swinging-bucket rotor)
No. per Case60
No. per Pack1
Quantity100/Cs.
Shelf Life5 Years after month of production
Temperature Range (Metric)-20°C to +60°C
Unit SizeCase