CELLSTAR 96-well, Cell Culture-Treated, Flat-Bottom Microplate
Actual product may vary
Greiner Bio-One
CELLSTAR 96-well, Cell Culture-Treated, Flat-Bottom Microplate
Color:White; Sterility:Sterile; Lid:With Lid
Have Questions?
Catalog NumberColor
FIS07-000-138White
Catalog number FIS07-000-138
Price (SGD)
381.00
Case
Color:
White
Price (SGD)
381.00
Case
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Compatible with automated systems
  • Alphanumeric well coding
  • High clarity of microplates for optimal microscopic examinations
  • Stackable
  • Individu
Specifications
LidWith Lid
Array8 x 12
AutoclavableNot Autoclavable
ColorWhite
MaterialPolystyrene
No. of Wells96
Volume (Metric) Working25 to 340 μL
SterilitySterile
Surface TreatmentCell Culture
Total Volume (Metric) Well392 μL
Well ShapeFlat Bottom (F-well)
Max. RCF4800 x g (swinging bucket rotor)
No. per Case60
No. per Pack8
Quantity32/Cs.
Temperature Range (Metric)-20°C to +60°C
Well DesignChimney
Unit SizeCase