LUMITRAC™ and FLUOTRAC™ Binding 96-well Polystyrene Microplates
Actual product may vary
Greiner Bio-One
LUMITRAC™ and FLUOTRAC™ Binding 96-well Polystyrene Microplates
For luminescence measurements. Greiner Bio-One LUMITRAC™ and FLUOTRAC™ Binding 96-Well Polystyrene Microplates are specifically treated to provide an increased protein binding.
Have Questions?
Catalog NumberColorSurface TreatmentSterility
FIS07-000-130WhiteLUMITRAC 200Non-sterile
Catalog number FIS07-000-130
Price (SGD)
272.00
Case
Color:
White
Surface Treatment:
LUMITRAC 200
Sterility:
Non-sterile
Price (SGD)
272.00
Case
  • Non-Pyrogenic
  • Material: Polystyrene
  • Free of detectable DNase, RNase, human DNA
  • Traced way back to production through defined LOT number system
  • Sample carryover and cross-contamination risk is minimized
Specifications
FormatStandard
LidWithout Lid
Array8 x 12
Binding TypeMedium Binding
ColorWhite
MaterialPolystyrene
No. of Wells96
SterilityNon-sterile
Surface TreatmentLUMITRAC 200
Well ShapeFlat Bottom (F-well)
Quantity40/Cs.
Well DesignChimney
Unit SizeCase