96-well Non-treated Polystyrene Microplates
Actual product may vary
Greiner Bio-One
96-well Non-treated Polystyrene Microplates
For agglutination tests. Greiner Bio-One 96-Well Non-treated Polystyrene Microplates is free of detectable DNase, RNase, human DNA.
Have Questions?
Catalog NumberWell ShapeSterilityQuantity
FIS07-000-124Flat Bottom (F-well)Non-sterile100/Cs.
Catalog number FIS07-000-124
Price (SGD)
291.00
Case
Well Shape:
Flat Bottom (F-well)
Sterility:
Non-sterile
Quantity:
100/Cs.
Price (SGD)
291.00
Case
  • Non-Pyrogenic
  • Free of detectable DNase, RNase, human DNA
  • Material: Polystyrene
  • Can be traced all the way back to production through a defined LOT number system
  • No sharp corners, can be pipetted easily and cleanly
Specifications
FormatStandard
LidWithout Lid
Array8 x 12
ColorClear
MaterialPolystyrene
No. of Wells96
SterilityNon-sterile
Surface TreatmentNon-treated
Total Volume (Metric) Well382 μL (Working volume: 25-340μL)
Well ShapeFlat Bottom (F-well)
No. per Case100
Quantity100/Cs.
Unit SizeCase