▸
Microplates
▸
Storage Microplates
▸
96-well Non-treated Polypropylene Microplates
Actual product may vary
Greiner Bio-One
96-well Non-treated Polypropylene Microplates
Features high chemical resistance. Greiner Bio-One 96-Well Non-treated Polypropylene Microplates is free of detectable DNase, Rnase and human DNA.
Have Questions?
Change view
Catalog Number
Color
Sterility
Well Shape
FIS07-000-110
Black
Non-sterile
Flat Bottom (F-well)
FIS07-000-153
Clear
Non-sterile
Conical Bottom (V-well)
2 Options
Catalog number
FIS07-000-110
Price (SGD)
737.00
Case
Add to cart
Color:
Black
Sterility:
Non-sterile
Well Shape:
Flat Bottom (F-well)
Price (SGD)
737.00
Case
Add to cart
Non-Pyrogenic
Free of detectable DNase, RNase, human DNA
Material: Polypropylene
High chemical resistance
Sealable with adhesive films and heat sealer
Can be traced back to production through defined LOT nu
Specifications
Format
Standard
Lid
Without Lid
Array
8 x 12
Color
Black
Material
Polypropylene
No. of Wells
96
Sterility
Non-sterile
Surface Treatment
Non-treated
Well Shape
Flat Bottom (F-well)
Quantity
100/Cs.
Well Design
Chimney