384-well Small Volume™ HiBase Polystyrene Microplates
Actual product may vary
Greiner Bio-One
384-well Small Volume™ HiBase Polystyrene Microplates
Features round wells with a conical geometry. Greiner Bio-One 384-Well Small Volume™ HiBase Polystyrene Microplates is free of detectable DNase, RNase, human DNA.
Have Questions?
Catalog NumberColorSterilityNo. per Case
FIS07-000-066WhiteNon-sterile40
Catalog number FIS07-000-066
Price (SGD)
1,212.00
Case
Color:
White
Sterility:
Non-sterile
No. per Case:
40
Price (SGD)
1,212.00
Case
  • Non-Pyrogenic
  • Material: Polystyrene
  • Free of detectable DNase, RNase, human DNA
  • Traced way back to production through defined LOT number system
  • Working volume: 4μL - 25μL
Specifications
FormatStandard
LidWithout Lid
DescriptionPlate design: HiBase
Array16 x 24
Binding TypeMedium Binding
ColorWhite
MaterialPolystyrene
No. of Wells384
SterilityNon-sterile
No. per Case40
Unit SizeCase