Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates
Actual product may vary
Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates
Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates
Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates
Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates
Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates
Store samples safely with these low-binding 96-well polypropylene plates, which will not allow DNA or proteins to bind and will allow for complete sample recovery.
Have Questions?
Catalog NumberColorSterilityWell ShapeFit for Purpose
249944NaturalNon-sterileRound, Conical Bottom (V-well)Certified Quality
Catalog number 249944
HSN 39269099
Price (SGD)
23.70
Special offer
Ends: 30-Nov-2026
79.00
Save 55.30 (70%)
PK
Color:
Natural
Sterility:
Non-sterile
Well Shape:
Round, Conical Bottom (V-well)
Fit for Purpose:
Certified Quality
Price (SGD)
23.70
Special offer
Ends: 30-Nov-2026
79.00
Save 55.30 (70%)
PK
Achieve complete sample recovery with Thermo ScientificTM NuncTM 96-Well Polypropylene MicroWellTM Plates, which are the ideal storage plate as polypropylene has a lower binding capacity so proteins or DNA will not bind. Withstands temperatures from -80° to +121°C. Available in a variety of colors for quick identification of storage plates.
Features:
  • Shared wall design increases the well volume
  • Wider wells and rounded bottom improve mixing
  • Conical bottom well for optimal sample recovery
  • Black and white plates available for fluorescent- and luminescent-based applications
  • Low binding polypropylene for homogeneous assays and storage
Specifications
BarcodeCustom Barcoding Available
Certifications/ComplianceCertified RNase and DNase Free. ANSI Compliant.
FormatStandard
LidWithout Lid
Description96-well Polypropylene Sample Processing & Storage Plates
Array8 x 12
AutoclavableAutoclavable
Packaging20 pk., 120 cs.
Binding TypeLow binding PP
Product LineNunc
ColorNatural
Product Type96-well plate
For Use With (Equipment)ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
MaterialPolypropylene
Volume (Metric) Well450 μL
No. of Wells96
Volume (Metric) Working400 μL
SterilityNon-sterile
Surface TreatmentNon-treated
Well ShapeRound, Conical Bottom (V-well)
Dnase FreeYes
For Use With (Application)Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
Green FeaturesACT Ecolabel (Gold)
No. per Case120
No. per Pack20
Quantity120/Cs.
Regulatory StatusRUO
RNase FreeYes
Fit for PurposeCertified Quality
Plate SizeANSI/ SLAS Standard
Unit SizePK