Acid Digestion Bomb Replacement Cups
Acid Digestion Bomb Replacement Cups
Actual product may vary
Acid Digestion Bomb Replacement Cups
For use with Parr General Purpose Acid Digestion Bombs
Have Questions?
Catalog NumberFor Use With (Equipment)
PARA280AC24744 general purpose acid digestion bomb
Catalog number PARA280AC2
Price (SGD)
640.00
EA
Add to cart
For Use With (Equipment):
4744 general purpose acid digestion bomb
Price (SGD)
640.00
EA
Add to cart
Specifications
Product TypeCup with Cover
Capacity (English)0.77 oz.
Capacity (Metric)23 mL
For Use With (Application)4744 general purpose acid digestion bomb
For Use With (Equipment)4744 general purpose acid digestion bomb
MaterialPTFE
Unit SizeEA