96-Well, Cell Culture-Treated, U-Shaped-Bottom Microplate
96-Well, Cell Culture-Treated, U-Shaped-Bottom Microplate
Actual product may vary
96-Well, Cell Culture-Treated, U-Shaped-Bottom Microplate
96-Well, Cell Culture-Treated, U-Shaped-Bottom Microplate
96-Well, Cell Culture-Treated, U-Shaped-Bottom Microplate
96-Well, Cell Culture-Treated, U-Shaped-Bottom Microplate
96-Well, Cell Culture-Treated, U-Shaped-Bottom Microplate
Color:Clear; Sterility:Sterile; Lid:With Lid
Have Questions?
Catalog NumberColor
LSVFB012932Clear
Catalog number LSVFB012932
Price (SGD)
144.00
Special offer
Ends: 31-Mar-2026
189.00
Save 45.00 (24%)
Case
Add to cart
Color:
Clear
Price (SGD)
144.00
Special offer
Ends: 31-Mar-2026
189.00
Save 45.00 (24%)
Case
Add to cart
  • Uniform well volume to ensure equal growth surface area
  • Well surface smooth, free from striation to maximize usable growth area
  • Raised rims on wells with uniform rings on lid to reduce evaporation
  • Single position lid reduces risks of cross-contamination, handling mistakes
  • Alphanumeric code on wells for easy identification
  • Suitable for all common instruments, automation
  • Sterilized by gamma irradiation
  • Non-pyrogenic
Specifications
BarcodeAlphanumeric
Certifications/ComplianceCE
FormatStandard
LidWith Lid
Culture Area0.32 cm2
Array8 x 12
Packaging1/Pk., 100/Cs.
ColorClear
Product TypeTissue Culture Plate
MaterialVirgin Polystyrene
Volume (Metric) Well0.32 mL
No. of Wells96
SterilitySterile
Surface TreatmentCell Culture
Well ShapeRound Bottom (U-well)
For Use With (Application)Tissue culture
No. per Pack100 Plates
RimmedYes
Well DesignRaised rims with condensation rings
Unit SizeCase