▸
Flasks
▸
Freeze Drying Flasks
▸
Fast-Freeze™ Flasks, Complete Assembly
Actual product may vary
Labconco
Fast-Freeze™ Flasks, Complete Assembly
Fast-Freeze™ Flasks are easy to handle and fast to load.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
LBC7540600
300 mL
LBC7540000
40 mL
FIS10-269-53
300 mL
LBC7540800
600 mL
LBC7541000
1200 mL
LBC7541200
2000 mL
LBC7540300
120 mL
LBC7540900
900 mL
LBC7541100
750 mL
9 Options
Catalog number
LBC7540600
Price (SGD)
611.00
Each
Add to cart
Capacity (Metric):
300 mL
Price (SGD)
611.00
Each
Add to cart
Easy to handle and fast to load
Flat borosilicate glass
Silicone rubber snap-on tops provide reliable high-vacuum seals
Eliminate risk of contamination by vacuum grease; reduce spillage and sample loss
No easily misplaced small parts
Wide openings
Should be filled no more than one-third of the volume so a maximum surface area is achieved
Compatible with all major brands of freeze-drying equipment, including Labconco and Virtis™
Specifications
Diameter (English) Outer
2.32 in.
Diameter (Metric) Flask Top Adapter
1.9 cm
Capacity (Metric)
300 mL
Material
Borosilicate Glass
For Use With (Equipment)
Compatible with all major brands of freeze-drying equipment
Height (English)
5.7 in.
Height (Metric)
14.5 cm
Product Type
Flask
Weight (English)
1.5 lb.
Disposable
Reusable
Mouth Style
Wide
Bottom Shape
Flat
Diameter (Metric) Outer
59.2 mm
Weight (Metric)
0.7 kg