Kimtech™ Purple Nitrile™ Gloves
Actual product may vary
Kimtech™ Purple Nitrile™ Gloves
Purple nitrile glove offers maximum protection for higher-risk procedures. Unique color differentiates from latex and other synthetic gloves.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
KLC55084X-Large
KLC55080X-Small
FIS19-149-863CLarge
KLC55083Large
FIS19-149-863DX-Large
Catalog number KLC55084
Price (SGD)
-
Size:
X-Large
Purple nitrile glove offers maximum protection for higher-risk procedures. Unique color differentiates from latex and other synthetic gloves.
  • Textured fingertips offers dexterity and tactile sensitivity
  • Latex-free
  • Ambidextrous
  • 9.5 in. length
  • Beaded cuff prevent roll-down and allow for easy donning
  • Powder free
Specifications
Certifications/ComplianceASTM D6978-05 (non-sterile only), Meets USP <800> guidelines
Chemotherapeutic RatedYes
ApplicationsLaboratory, Healthcare
AQL0.65
ColorPurple
DisposableSingle-use
Exterior FinishPartially Textured
Finger Thickness (English)6 mil
Finger Thickness (Metric)0.15 mm
Latex FreeYes
MaterialNitrile
Palm Thickness (English)4.7 mil
Palm Thickness (Metric)0.12 mm
PowderedPowder Free
SizeX-Large
Thickness (English)6 mil
Thickness (Metric)0.15 mm
FDA 510 (k)Yes
Model55084
SeriesKimtech™
Tensile Strength21 MPa
Length (English)9.5 in.
Length (Metric)24.13 cm
Unit SizePK