Kimtech™ Sterling™ Nitrile-XTRA 12 in. Gloves
Actual product may vary
Kimtech™ Sterling™ Nitrile-XTRA 12 in. Gloves
Sterling™ Nitrile-XTRA gloves are 12 in. in length for added protection and combine the sensitivity of latex with the strength of nitrile.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
KLC53137X-Small
FIS19-050-591Small
KLC53138Small
FIS19-050-592Medium
KLC53139Medium
KLC53140Large
Catalog number KLC53137
Price (SGD)
320.00
Special offer
Ends: 06-Sep-2026
383.00
Save 63.00 (16%)
Case
Size:
X-Small
Price (SGD)
320.00
Special offer
Ends: 06-Sep-2026
383.00
Save 63.00 (16%)
Case
Sterling™ Nitrile-XTRA gloves are 12 in. in length for added protection and combine the sensitivity of latex with the strength of nitrile.
  • XTRA long 12 in. length
  • Powder free
  • non-sterile
  • Static dissipative
  • Textured fingertips
  • Beaded cuff
  • Ambidextrous
  • Typical uses include: Laboratory, Cleaning, Maintenance, Electronic Assembly and Inspection
Specifications
Certifications/ComplianceASTM D 6319, ASTM D6978-05, Meets NFPA standard 1999:2013, Meets USP <800> guidelines
Chemotherapeutic RatedYes
ChlorinatedYes
ApplicationsHealthcare, Laboratory, Industrial
AQL1
ColorGray
DisposableSingle-use
Exterior FinishPartially Textured
Finger Thickness (English)3.5 mil
Finger Thickness (Metric)0.08 mm
Latex FreeYes
MaterialNitrile
Palm Thickness (English)3.1 mil
Palm Thickness (Metric)0.07 mm
PowderedPowder Free
Product TypeNitrile Examination Glove
SizeX-Small
SterilityNon-sterile
Thickness (English)3.5 mil
Thickness (Metric)0.08 mm
FDA 510 (k)Yes
Glove Cuff StyleBeaded
Model53137
Quantity1000/Cs.
SeriesKimtech™
Tensile Strength42 MPa
Length (English)12 in.
Length (Metric)30.5 cm
Unit SizeCase